Learn More
Cytiva Whatman™ Grade 589/2 Quantitative Filter Paper
Retains medium fine precipitates using material with virtually no ash content. Whatman™ Grade 589/2 Quantitative Filter Paper is used when medium fine precipitates are being collected and papers must be ashed to support analysis.
240.00 NOK - 1820.00 NOK
Specifications
Color Code | White |
---|---|
Material | Cotton Linters |
Particle Retention | 4 to 12 μm |
Filtration Speed | 70 s |
Flow Rate | Medium |
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
11355194
|
Cytiva
10300106 |
50 mm | Disc |
248.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11365194
|
Cytiva
10300107 |
55 mm | Disc |
256.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11315204
|
Cytiva
10300112 |
150 mm | Disc |
770.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10028030
|
Cytiva
10300110 |
110 mm | Disc |
473.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11335204
|
Cytiva
10300120 |
240 mm | Disc |
1820.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11345204
|
Cytiva
10300143 |
110 mm | Folded |
775.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10619991
|
Cytiva
10300102 |
12.7 mm | Disc |
600.00 NOK
Pack of 1000 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11305204
|
Cytiva
10300111 |
125 mm | Disc |
680.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11345194
|
Cytiva
10300103 |
40.5 mm | Folded |
240.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10262181
|
Cytiva
10300108 |
70 mm | Folded |
305.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
10727371
|
Cytiva
10300109 |
90 mm | Folded |
422.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11325204
|
Cytiva
10300114 |
185 mm | Folded |
1165.00 NOK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
Description
Retains medium fine precipitates using material with virtually no ash content. Whatman™ Grade 589/2 Quantitative Filter Paper is used when medium fine precipitates are being collected and papers must be ashed to support analysis.
Designed for gravimetric analysis- White Ribbon Filter for medium fine precipitates
- Ashless
- Medium filtration speed
- For sand content measurement in foodstuffs
- For flour grade determination
- For aqueous suspension analysis
Specifications
White | |
4 to 12 μm | |
Medium | |
Ashless |
Cotton Linters | |
70 s | |
180 μm |