Learn More
Cytiva Whatman™ Grade 20 Chr Cellulose Chromatography Paper
Product Code. 11378854
Change view
Click to view available options
Length (Metric):
46 cm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11378854 46 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11378854 Supplier Cytiva Supplier No. 3020917
  • ONLINE EXCLUSIVE PRICE
    4755.00 NOK
    4332.05 NOK / Pack of 100
    Save 422.95 NOK
    8% Off
Estimated Shipment: 02-04-2027
Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind
TRUSTED_SUSTAINABILITY
Specifications
Description 20 Chr cellulose chromatography sheets is a 0.17 mm paper for separation of samples of unknown composition, with outstanding resolution (at low loadings.)
Dimensions (L x W) 46 x 57 cm (18.1 x 22.4 in.)
Film Thickness 0.17 mm
Finish Smooth
For Use With (Application) For Optical or Radiometric Scanning and Also Used in Protein and Nucleic Acid Blotting
Format Sheet
Grade Grade 20 Chr
Length (English) 18.11 in.
Material Cellulose
Width (English) 22.4 in.
Width (Metric) 57 cm
Flow Rate 85 mm/30 min.
Length (Metric) 46 cm