Learn More
Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper
Product Code. 10622245
Change view
Click to view available options
Length (Metric):
22 cm
30 cm
57 cm
Width (Metric):
2.5 cm
46 cm
Format:
Roll
Sheet
Unit Size:
Each
Pack of 100
Product Code. Length (Metric) Width (Metric) Format unitSize
10622245 22 cm 2.5 cm Sheet Pack of 100
11909197 30 cm 2.5 cm Roll Each
11308854 57 cm 46 cm Sheet Pack of 100
3 options
Product Code. 10622245
Brand: Cytiva 30178793
Only null left
Add to basket
Add to basket
17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
TRUSTED_SUSTAINABILITY
Specifications
Description 17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
Dimensions (L x W) 2.5 x 22 cm (0.98 x 8.66 in.)
Film Thickness 0.92 mm
For Use With (Application) Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Format Sheet
Grade Grade 17 Chr
Length (English) 8.66 in.
Material Cellulose
Width (English) 0.98 in.
Width (Metric) 2.5 cm
Flow Rate 190 mm/30 min.
Length (Metric) 22 cm