Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ FF80HP Membranes
Shop All Cytiva Products
Click to view available options
:
FF80HP Membrane
Description
- Uniform surface without unincorporated nitrocellulose powder
- A fine structure fiber distribution provides large internal surfaces for binding proteins
- Exceptional intra- and inter-lot consistency with a coefficient of variation (CV) for capillary rise of < 10%
- Improved assay consistency and reproducible limit of detection
- Reduced assay optimization costs, compared to a less consistent membrane
Specifications
Specifications
| Material | Polyester Backing |
| Material (Membrane) | Nitrocellulose |
| Flow Rate | 60 to 100 s/4 cm |
| For Use With (Application) | High Density Samples in a Lateral Flow Assay |
| Includes | 100 μm Backing |
| No. per Pack | 10/Pk. |
| Thickness (Metric) | 200 μm |
| Type | FF80HP Membrane |
| Length (Metric) | 297 mm |
| Width (Metric) | 210 mm |