Learn More
Cytiva Whatman Electronic Butler
The Whatman eButler rapid membrane dispenser allows for more efficient microbiological testing.
Brand: Cytiva 10477103
25707.00 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 3.10500kg
Description
- Aluminum and stainless-steel construction
- Designed for easy membrane loading and device usability
- Both push-button and touch-free operation modes to suit your preference
- Up to 400 membranes per package
- Supports rapid dispensing of the culture membrane
- Compatible with Whatman sterile mixed cellulose ester (MCE) microbiology membranes
Specifications
Hands-free or push button operation | |
Aluminum, Stainless Steel | |
Dispensing culture membranes | |
Electronic Butler |