Learn More
QIAGEN EasiCollect
missing translation for 'orderingAttributeHoverText'
Quantity:
50 Pk.
missing translation for 'unitSize'
Pack of 50
Description
For simple, reliable and standardized sample collection.
50 buccal sample collection devices for simplifies sample uptake and transportation.
Specifications
Specifications
| Sample Type | Buccal Cells, Saliva |
| Includes | Includes an Indicating FTA Card that changes color from pink to white upon sample addition |
| Product Type | DNA Sample Collection Device |
| Quantity | 50 Pk. |
Safety and Handling
Signal Word
- Warning