Learn More
- Home
- Products
- Filters and Filtration
- Filtration Membrane Discs
- Cytiva Whatman™ Cellulose Acetate Membranes
Cytiva Whatman™ Cellulose Acetate Membranes
Product Code. 11359024
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.8 μm
1.2 μm
Diameter (Metric):
13 mm
25 mm
47 mm
50 mm
85 mm
110 mm
142 mm
293 mm
Membrane Type:
OE 66
OE 67
ST 68
ST 69
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Pack of 50
| Product Code. | Pore Size | Diameter (Metric) | Membrane Type | Sterility | unitSize |
|---|---|---|---|---|---|
| 11359024 | 0.45 μm | 85 mm | OE 67 | Non-sterile | Pack of 50 |
| 11385344 | 0.8 μm | 47 mm | ST 68 | Non-sterile | Pack of 100 |
| 11313425 | 1.2 μm | 47 mm | ST 69 | Non-sterile | Pack of 100 |
| 11329024 | 0.45 μm | 25 mm | OE 67 | Non-sterile | Pack of 100 |
| 11309024 | 0.45 μm | 13 mm | OE 67 | Non-sterile | Pack of 100 |
| 11349024 | 0.45 μm | 50 mm | OE 67 | Non-sterile | Pack of 100 |
| 11399014 | 0.45 μm | 110 mm | OE 67 | Non-sterile | Pack of 50 |
| 11319024 | 0.45 μm | 142 mm | OE 67 | Non-sterile | Pack of 25 |
| 11335354 | 0.2 μm | 47 mm | OE 66 | Non-sterile | Pack of 100 |
| 11345354 | 0.2 μm | 50 mm | OE 66 | Non-sterile | Pack of 100 |
| 11355354 | 0.2 μm | 110 mm | OE 66 | Non-sterile | Pack of 50 |
| 11375354 | 0.2 μm | 293 mm | OE 66 | Non-sterile | Pack of 25 |
| 11385354 | 0.2 μm | 47 mm | OE 66 | Sterile | Pack of 100 |
| 11344814 | 0.45 μm | 25 mm | ST 69 | Non-sterile | Pack of 100 |
| 11364814 | 0.2 μm | 47 mm | ST 69 | Non-sterile | Pack of 100 |
| 10265591 | 0.45 μm | 47 mm | ST 69 | Non-sterile | Pack of 100 |
| 11325354 | 0.2 μm | 25 mm | OE 66 | Non-sterile | Pack of 100 |
| 11339024 | 0.45 μm | 47 mm | OE 67 | Non-sterile | Pack of 100 |
| 11365354 | 0.2 μm | 142 mm | OE 66 | Non-sterile | Pack of 25 |
| 11395354 | 0.2 μm | - | OE 66 | Non-sterile | Pack of 5 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
- Low protein binding capacity
- Hydrophilic
- Improved solvent resistance and heat resistance
- Can be used for liquids or gases up to 180°C
- Sterilizable by any method without sacrificing integrity
For filtration with aqueous or alcoholic solvents and gases
Specifications
Specifications
| Material | Cellulose Acetate |
| Pore Size | 0.45 μm |
| Sterility | Non-sterile |
| Flow Rate | 12s/100mL/12.5cm2 |
| Hydrophobicity | Hydrophilic |
| Membrane Type | OE 67 |
| No. per Pack | 50/Pk. |
| Diameter (Metric) | 85 mm |