Learn More
QIAGEN QIAcard Bloodstain
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
Description
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
- Non-Indicating and Non-FTA treated card designed for the collection, short-term handling of specimens.
- Unlike FTA Cards, the Bloodstain Cards are not coated with a protective chemical cocktail and DNA is as risk of degradation and damage.
- Convenient transport of blood and bodily fluids with protective cover included.
Specifications
Specifications
| Format | Card |
| For Use With (Application) | Collection and transport of blood and body fluids |
| Quantity | 100 Pk. |
| Sample Volume | 125μL per sample area |