Learn More
Cytiva Whatman™ Application Specific Filters
Acid-washed filter for the determination of trace elements (Mg, Mn, Co, Cu, Mo, B).
Brand: Cytiva 10310643
617.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.09000kg
Description
Whatman offers a line of cellulose filter papers for specific applications. The product line includes filter papers for use in soil analysis and filter papers for the sugar industry.Specifications
For use in soil analysis, filter papers for sugar industry | |
Grade 512.5 | |
110mm | |
Filter |