Learn More
Cytiva Whatman™ Acid/Alkali Test Papers
Product Code. p-194521
Change view
Click to view available options
Color Indicator:
Litmus Blue
Litmus Red
Format:
Reel
Strip
Unit Size:
Each
Pack of 10
Product Code. Color Indicator Format unitSize
11894463 Litmus Blue Strip Pack of 10
11390674 Litmus Red Reel Each
11804473 Litmus Red Strip Pack of 10
3 options
Product Code. 11894463
Brand: Cytiva 2600601
Only null left
Add to basket
Add to basket
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3
Order Info
20 leaves per book. 10 x 10 books per pack.
TRUSTED_SUSTAINABILITY
Specifications
Color Indicator Litmus Blue
pH Range 5 to 8
Format Strip
Quantity 10 x 10 Books of 20 Strips
Type pH Test Paper
Length (Metric) 60 mm
Width (Metric) 10 mm