Learn More
Cytiva Whatman™ 90mm Nuclepore™ Polycarbonate Track-Etched Membranes
Product Code. 11378984
Change view
Click to view available options
Pore Size:
0.05 μm
0.1 μm
0.2 μm
0.4 μm
1 μm
2 μm
3 μm
Unit Size:
Pack of 25
Product Code. Pore Size unitSize
11378984 0.1 μm Pack of 25
11368984 0.05 μm Pack of 25
11332915 0.2 μm Pack of 25
11388984 0.4 μm Pack of 25
11398984 1 μm Pack of 25
11308994 2 μm Pack of 25
11318994 3 μm Pack of 25
7 options
Product Code. 11378984
Brand: Cytiva 111705
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
TRUSTED_SUSTAINABILITY
Specifications
Material Nucleopore Polycarbonate
Pore Size 0.1 μm
Flow Rate High
For Use With (Application) Epifluorescence microscopy, environmental analysis, cell biology
Hydrophobicity Hydrophilic
Diameter (Metric) 90 mm