Learn More
Cytiva Whatman™ 903 Proteinsaver Snap-Apart Card
Description
903 sample collection card with 4 sample spots.
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with 4 half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75-80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
- Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
- Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
Specifications
Specifications
| Material | Cellulose |
| For Use With (Application) | Sample collection, transport, and archiving |
| Includes | 4 Sample Spots |
| Product Line | Whatman |
| Type | 903 Proteinsaver Snap-Apart Card |
| Volume (Metric) | 4 x 75 to 80μL |