Learn More
Cytiva Whatman™ 850-DS 8-Channel Filter Plate
Use with Agilent™ 850-DS Dissolution Sampling Station for automated sample preparation in dissolution testing
Brand: Cytiva 7707-3900
8326.75 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 1.70000kg
Description
- Exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station
- Simplifies filter replacement between timepoints
- Reliable alignment of the liquid path increases productivity by reducing the risk of jamming, and reducing leaks that may occur with other dissolution sample preparation systems
- Save time and eliminate errors associated with manual sampling
- Automated processing of up to 8 samples simultaneously
- Readily available in a wide range of pore sizes and materials
Specifications
Agilent 850-DS Dissolution Sampling Station | |
Automated Sample Preparation in Dissolution Testing | |
8 Channel |
Glass Microfiber | |
50/Pk. |