missing translation for 'onlineSavingsMsg'
Learn More

Proteintech WFDC3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24917-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893425

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains four WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Alternatively spliced transcript variants have been observed but their full-length nature has not been determined.
Specifications

Specifications

WFDC3
Polyclonal
Unconjugated
WFDC3
WAP14, WFDC3
Rabbit
Antigen Affinity Chromatography
RUO
140686
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IUB2
WFDC3
WFDC3 Fusion Protein Ag20207
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers