Learn More
Thermo Scientific™ Well Plate Accessories
Product Code. 15192799
Change view
Click to view available options
Product Type:
Automated Sample Concentrator
Center Hole Cap
Deep Well Plate Vial
MTRS Vial
Manual Concentrator
Mat Applicator
Microtiter Plate Vial
Plug
Sealing Tape Applicator
Square Well Plate Vial
Tube Tray
Volume (Metric) Working:
100 μL
1000 μL
1100 μL
1500 μL
350 μL
500 μL
700 μL
800 μL
Not Applicable
Well Shape:
Flat Bottom (F-well)
Not Applicable
Round
Square
Taper
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 2
Pack of 500
Pack of 96
Pack of 960
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Volume (Metric) Working Well Shape Unit Size
15192799 Center Hole Cap Not Applicable Not Applicable Pack of 96
15152799 Microtiter Plate Vial 100 μL Flat Bottom (F-well) Pack of 960
15162799 Deep Well Plate Vial 350 μL Taper Pack of 960
15122779 Microtiter Plate Vial 500 μL Taper Pack of 500
15142779 Microtiter Plate Vial 700 μL Round Pack of 100
15132779 Microtiter Plate Vial 700 μL Taper Pack of 500
15182799 MTRS Vial 800 μL Round Pack of 960
15182779 Square Well Plate Vial 1000 μL Square Pack of 960
15172799 Square Well Plate Vial 1000 μL Round Pack of 960
15152779 Microtiter Plate Vial 1100 μL Round Pack of 500
15162779 Deep Well Plate Vial 1100 μL Round Pack of 500
15172779 Deep Well Plate Vial 1500 μL Round Pack of 500
15182719 Mat Applicator Not Applicable Not Applicable Each
15172719 Sealing Tape Applicator Not Applicable Not Applicable Pack of 2
15142809 Tube Tray Not Applicable Not Applicable Pack of 10
15102809 Plug Not Applicable Not Applicable Pack of 96
15152809 Manual Concentrator Not Applicable Not Applicable Each
15162809 Automated Sample Concentrator Not Applicable Not Applicable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 15192799 Supplier Thermo Scientific™ Supplier No. 60180C100
 Restricted Item
Estimated Shipment: 12-10-2026
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.
Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.
Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Webseal
Color Red
Product Type Center Hole Cap
For Use With (Equipment) 96-well Plate Inserts
Material Silicone, PTFE
No. of Wells 96
Volume (Metric) Working Not Applicable
Well Shape Not Applicable
Includes Septum – Silicone/PTFE, Pre-Slit, 45°
Plate Compatibility 96-well Plates
What is the difference between the SureSTART Performance Level 1, Performance Level 2, and Performance Level 3 vials?
All glass vials found within the three different performance levels are made from Type 1 Class A Borosilicate glass. This type of glass has excellent chemical resistance properties that make it ideal for the containment or storage of acidic, neutral and alkaline samples/analytes. The vials also have high temperature resistance and are suitable for use at temperatures between -50 degrees C to 120 degrees C (although a rapid change in temperature of the products should be avoided).
The Performance Level 1 vials are designed to identify those which are suitable for simple routine, testing with limited budget. Performance Level 1 glass vials have a single wavy line.
For most analytical requirements, the Performance Level 2 products will be suitable for use with higher sensitivity methods while maintaining throughput. Performance Level 2 glass vials have two wavy lines.
For the most stringent analytical requirements, the Performance Level 3 products have advanced features which make them suitable for use with difficult samples in terms of chemistry and available sample volume. All Performance Level 3 products are made from 33-expansion glass and these vials have three wavy lines.
What happened to National, Chromacol, and other brands for chromatogenic workflow accessories?
Many of the products that were branded as National Scientific and Chromacol have been absorbed into the new Thermo Scientific SureSTART Collection, although there are still a few products that still exist within the marketplace.
For more information, please contact your local sales representative or complete the inquiries form at the bottom of the SureSTART Autosampler Vials and Caps page (https://www.thermofisher.com/us/en/home/industrial/chromatography/chromatography-consumables/autosampler-vials-caps-hplc-gc.html).
What is the SureSTART products portfolio for chromatographic workflows?
The SureSTART portfolio is a comprehensive list of vials, caps, inserts, well plates, mats and accessories that are suitable for use with chromatographic workflows. The SureSTART portfolio contains three different performance levels that have been quality checked to provide a robust and reliable product.
The SureSTART portfolio includes products that were previously found in our legacy brands such as National Scientific, Chromacol, and Sun-Sri.