Learn More
SI Analytics™ Weighing Funnel
Product Code. 10631024 Shop All SI Analytics Products
Change view
Click to view available options
Volume (Metric):
3 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10631024 3 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10631024 Supplier SI Analytics™ Supplier No. 285221130
  • ONLINE EXCLUSIVE PRICE
    476.00 NOK
    431.71 NOK / Pack of 1
    Save 44.29 NOK
    9% Off
 Restricted Item
Estimated Shipment: 01-12-2026
SI Analytics™ Weighing Funnel is an accessory for use with TitroLine™ and Titronic™ models.
  • Glass funnel
  • Holds up to 3 mL
TRUSTED_SUSTAINABILITY
Specifications
Type Weighing Funnel
Material Glass
Volume (Metric) 3 mL