Learn More
Lenz Laborglasintrumente Weighing Bottles with NS Stopper
Product Code. p-7222638
Change view
Click to view available options
Capacity (Metric):
5 mL
70 mL
80 mL
Unit Size:
Pack of 6
Product Code. Capacity (Metric) unitSize
10559571 5 mL Pack of 6
15300885 80 mL Pack of 6
10028830 70 mL Pack of 6
3 options
Product Code. 10559571
Brand: Lenz Laborglasintrumente 05560002
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With interchangeable NS stopper
  • With inscription label, with fused-in size (mm) and capacity (ml)
  • Available in tall or low form
  • Stopper: DURAN™, bottle made of DURAN™ tubing
Specifications
Capacity (Metric) 5 mL