Learn More
Microspec™ Wedge Shaped Spreader
missing translation for 'orderingAttributeHoverText'
Quantity:
250
500
missing translation for 'unitSize'
Pack of 250
Pack of 500
Description
- Wedge Shaped Spreaders
- 37 mm x 180 mm,
- Sterile EtO
Specifications
Specifications
| Disposable | Single-use |
| Autoclavable | Not Autoclavable |
| Color | Blue |
| Packaging | Individually Wrapped in Microseal Pouch |
| Quantity | 250 |
| Size (English) | 1.46 x 7 in. |
| Size (Metric) | 37 x 180 mm |
| Material | Plastic |
| Product Type | Inoculating Spreader |
| Sterility | Sterile |