missing translation for 'onlineSavingsMsg'
Learn More

WDR92 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25540-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16874105

  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR92, also known as monad, contains two WD40 repeats is widely expressed in human tissues, especially testis. Overexpression of this protein or its binding partner RNA polymerase II-associated protein 3 (RPAP3) potentiated apoptosis and caspase-3 activation induced by TNF-a and cycloheximide, suggesting that WDR92, together with RPAP3, may function as a novel modulator of apoptosis.
Specifications

Specifications

WDR92
Polyclonal
Unconjugated
WDR92
WD repeat domain 92, WDR92
Rabbit
Antigen Affinity Chromatography
RUO
103784, 116143
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BGF3, Q96MX6
Wdr92
WDR92 Fusion Protein Ag22181
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers