missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27628-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Studies of the related mouse protein suggest that the encoded protein may play a role in the process of ossification.Specifications
| WDR8 | |
| Polyclonal | |
| Unconjugated | |
| WRAP73 | |
| WD repeat containing protein 8, WD repeat domain 8, WD repeat-containing protein WRAP73, WDR8, WRAP73 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 49856 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P2S5 | |
| WRAP73 | |
| WDR8 Fusion Protein Ag26267 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction