Learn More
WBSCR22 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28192-1-AP-150UL
Additional Details : Weight : 0.00020kg
Description
The WBSCR22 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human WBSCR22. This antibody recognizes human antigen. The WBSCR22 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
WBSCR22 | |
Polyclonal | |
Rabbit | |
Primary | |
Human |
Western Blot, Immunohistochemistry | |
Unconjugated | |
150 μL | |
114049 | |
IgG |