Learn More
Thermo Scientific™ Waterproof Cryo Gloves - Mid-Arm Length
Shop All Thermo Scientific Products
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
Each
Description
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
- Provides splash protection
- Comfortable, lightweight, breathable
- Side release buckles for quick on/off
Specifications
Specifications
| Description | Mid-Arm Length |
| Product Type | Waterproof Cryogenic Gloves |
| Size | Medium |
| For Use With (Application) | Cryogenic Preservation |
| Length (English) | 14 to 15 in. |
| Length (Metric) | 35.5 to 38 cm |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur