Learn More
Thermo Scientific™ WaterBugs™ Pseudomonas aeruginosa ATCC™ 27853™
Click to view available options
Quantity:
10 Test/Set
Unit Size:
Each
Description
Thermo Scientific™ WaterBugs™ Pseudomonas aeruginosa ATCC™ 27853™ is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
- Deliver <50 CFUs in a single inoculum to mimic water samples
- One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
Specifications
Specifications
| ATCC® | 27853TM*, 27853™* |
| Description | Pseudomonas aeruginosa ATCC™ 27853™ |
| Format | Quanti-Cult |
| Product Type | Quality Control Organism |
| Target Organism Class | Gram-negative Rods |
| Test Type | Quantitative QC |
| Packaging Type | 10 Test/Set |
| Quantity | 10 Test/Set |
| Strain Designations | Boston 41501 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.