Learn More
Thermo Scientific™ Water Softener Accessories
Product Code. 11396981
Change view
Click to view available options
Type:
Hardness Test Kit
Salt Tablets, 25 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11396981 Hardness Test Kit Each
11366971 Salt Tablets, 25 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11396981 Supplier Thermo Scientific™ Supplier No. 06.1000
  • ONLINE EXCLUSIVE PRICE
    440.00 NOK
    421.16 NOK / Pack of 1
    Save 18.84 NOK
    4% Off
View all promotions
In Stock
Add to basket
Add to basket
Remove hard ions, such as calcium and magnesium, using these accessories. Softened water extends the life of the reverse osmosis membrane.
Remove hard ions, such as calcium and magnesium, using Thermo Scientific™ Water Softener Accessories. Softened water extends the life of the reverse osmosis membrane.
Sensor controller for water softening monitoring
  • The Sensor controller SE is a monitoring system for a quality-controlled operation of the water softening system
Total hardness measurement (titration solution)
  • Necessary to determine the feed water hardness and calculate the softener program settings
Bacteria guard for water softeners
  • For disinfecting the ion exchange bed during each regeneration
  • The bacteria guard extracts chlorine from a part of the salt water to disinfect the ion exchange resin
Flexible connecting hose 1in. with stainless steel jacketing, length 1 meter
  • Smart and easy connection of your Thermo Scientific Water softener to wall connectors
  • Alternative instead of fixed piping, which offers more flexibility for maintenance
Mounting block 1in. with blending valve
  • For comfortable bypassing of the softener for maintenance, reparations, etc
Blending valve OVP for 1in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Blending valve OVP for 1 1/2in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cabinet Softening System WAE, Double Softening System WAT, Double Softening System WMS
Type Hardness Test Kit