Learn More
Si Analytics™ Water Distillation Units
Product Code. 10407742 Shop All SI Analytics Products
Change view
Click to view available options
Capacity (Metric):
4 L
7 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10407742 4 L Each
10098281 7 L Each
10098271 7 L Each
3 options
Product Code. 10407742
Brand: Si Analytics™ 285502028
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Incurre no costs for regeneration or replacement of resins, or for pretreatment modules that are required to protect the membrane. SI Analytics™ Water Distillation Unit is easy to clean and ensures maximum operating safety.
  • With the water distilling apparatus made of DURAN™ borosilicate glass 3.3 (hydrolytic class 1 glass), starting with normal tap water you can obtain a distillate of maximum purity that is substantially free from dissolved salts and of extremely low germ content.
  • A maximum conductivity of up to approx. 1μS/cm (25°C) is achieved in the distilled water.
  • The immersion heater is made of high-quality stainless steel, so that the presence of heavy-metal ions in the distillate is excluded.
  • The apparatus saves energy and water, because the cooling water that is heated up in the cooling coil is used for feed water.
  • The outflow of distillate is free from carbon dioxide because its temperature is 60°C.
  • Cost-effective in comparison with other water purification systems, such as ion exchangers or reverse osmosis apparatus
  • No costs are incurred for regeneration or replacement of resins, or for pretreatment modules that are required to protect the membrane.
  • The automatic water level control and built-in boil-dry protection ensure maximum operating safety.
  • Cleaning the apparatus is as easy as cleaning a household coffee maker.
  • In the standard version, the compactly built apparatus is supplied including the connecting cable, plug and hoses and is ready for connection.
  • Different versions are available.
TRUSTED_SUSTAINABILITY
Specifications
Description Water Distillation Unit, 4L, 230/14V/A Electrical input power
Diameter (Metric) Water Distillation Unit, 4L, 230/14V/A Electrical input power
Electrical Requirements 230 V 14 A
Voltage 230 V
Weight (Metric) 11 kg
Type Water Distillation Unit
Capacity (Metric) 4 L
Features Resistance Heating
Height (Metric) 750 mm
Length (Metric) 300 mm
Width (Metric) 300 mm