Learn More
Stuart™ Water Conversion Kit
Description
- Laboratories typically prefer to feed their water stills with deionized or pre-treated water to obviate the need for boiling and descaling
- This kit easily converts Aquatron Water Stills into a deionized water feed
- The still is then permitted to operate with most types of deionizer, and also piped supplies of pre-treated water
Specifications
Specifications
| For Use With (Application) | Conversion to Deionised Water Feed |
| For Use With (Equipment) | Aquatron Water Stills |
| Type | Water Conversion Kit |