Learn More
Water with 0.1% TFA, LC/MS grade, Thermo Scientific
Product Code. p-200109911 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Unit Size:
2.5 L
4 L
500mL
Product Code. 30280961
Brand: Thermo Scientific TL01190500
Request Bulk or Custom Format
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY
Specifications
Grade LC-MS
Quantity 500 mL
Form Liquid
Reagent Type Solvent Mixture
For Use With Liquid Chromatography, Mass Spectrometry