Learn More
Karl Hecht™ Watch Glasses
Description
- Acc. to DIN 12 341
- With fused rim
- Manufactured from glass
- Available in different diameters
Specifications
Specifications
| Description | Fused rim |
| Material | Glass |
| Shape | Round |
| Type | Watch Glass |
| For Use With (Application) | Evaporation, Heating, Holding |
| Diameter (Metric) | 10 cm |