Learn More
Hecht Karl™ Watch Glasses
15.79 NOK - 18.97 NOK
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10471702
|
Hecht Karl™
40010005 |
5 cm |
15.79 NOK
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
10237041
|
Hecht Karl™
40010008 |
8 cm |
16.91 NOK
Each |
In Stock
Log in to see stock. |
|||||
10738472
|
Hecht Karl™
40010007 |
7 cm |
17.38 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10329441
|
Hecht Karl™
40010009 |
9 cm |
18.97 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Specifications
Glass | |
Watch Glass |