Learn More
Gosselin™ Waste and Sharps Container, White Hinged Cap
Product Code. 15795542
Click to view available options
Capacity (Metric):
600 mL
800 mL
1.1 L
1.8 L
4.5 L
7 L
Quantity:
25/Cs.
42/Cs.
45/Cs.
60/Cs.
65/Cs.
70/Cs.
Unit Size:
Pack of 25
Pack of 42
Pack of 45
Pack of 60
Pack of 65
Pack of 70
Product Code. 15795542
Brand: Gosselin™ COX600A02
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CORNING GOSSELIN™ Waste and Sharps Container, White Hinged Cap is constructed of polypropylene and available in a variety of sizes.
  • Polypropylene construction
  • Suitable for waste and sharps
  • NFX 30-5111 compliant
  • Hinged cap
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 600 mL
Certifications/Compliance NFX 30-5111
Closure Type Hinged Top
Closure Color White
Color Yellow
Quantity 70/Cs.