Learn More
Duran™ Washing Bottle
Product Code. 10779671 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10779671 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10779671 Supplier Duran™ Supplier No. 247110109
  • ONLINE EXCLUSIVE PRICE
    750.00 NOK
    604.61 NOK / Pack of 1
    Save 145.39 NOK
    19% Off
In Stock
Manufactured from borosilicate glass
Manufactured from borosilicate glass
Specifications
Capacity (Metric) 100 mL
Type Gas Washing Bottle
Includes Bottom
Quantity 1 Each