Learn More
Thermo Scientific™ Wash Solution I (concentrated)
Product Code. 15319982
Click to view available options
Quantity:
360 mL
Unit Size:
360mL
Product Code. 15319982
Brand: Thermo Scientific™ R1611
Thermo Scientific GeneJET Wash Solution I (concentrated) is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491, K0492) and may be purchased separately.
Thermo Scientific GeneJET Wash Solution I (concentrated) is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491, K0492) and may be purchased separately. Provided in 360 mL aliquots.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Wash Buffer
Product Line GeneJET
Quantity 360 mL