Learn More
Thermo Scientific™ Wash Buffer, 1.0% Triton™ X-100, pH 8.0
Product Code. 15453199
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15453199
Brand: Thermo Scientific™ J60428.AK
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For GST-fusion protein purification
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer
Color Colorless
For Use With (Application) For GST-fusion protein purification
pH 8
Physical Form Liquid
Quantity 250 mL