Learn More
KARTELL Wash Bottle Screw Cap
Click to view available options
Closure Size:
DIN Std. GL18
DIN Std. GL32
Description
- Fit bottles part. no. 15881 - 15901
- Material: Polyethylene
Specifications
Closure Size | DIN Std. GL18 |
Product Type | Screw Cap |
Color | Translucent White |
Material | Polyethylene |
Quantity | 100 pcs. |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Disposable | Reusable |
Fit for Purpose | Certified Quality |