missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19755-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to WASF4.
WASF4, also named as SCAR2, belongs to the SCAR/WAVE family. WASF4 is a downstream effector molecule which involved in the transmission of signals from tyrosine kinase receptors and small GTPases to the actin cytoskeleton.Specifications
WASF4 | |
Polyclonal | |
Unconjugated | |
WASF4P | |
WASF4P | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
WAS protein family, member 4, WASF4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
644739 | |
-20°C | |
Liquid |