Learn More
Brady™ Bradylink™ Warning Chains
Product Code. 11751076
Change view
Click to view available options
Color:
Black/Yellow
Red/White
White
Yellow
Unit Size:
1 pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11751076 White 1 pack
10735715 Red/White 1 pack
11771076 Black/Yellow 1 pack
11781076 Yellow 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11751076 Supplier Brady™ Supplier No. 278268
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
  • Made of durable polyethylene, Bradylink™ Warning Chains are a decorative alternative to metal chains.
  • Chains are color coded to match Bradylink™ Warning Posts and have a special link design that prevents kinking.
  • Note: Bradylink Chains are not intended to support heavy objects and are recommended for indoor use only.
  • Package includes chain only (stanchions sold separately).
  • The Bradylink™ Warning Post System is non-conductive so it can be safely used in electrically hazardous areas.
Specifications
Product Type Warning Chain
Alternate Part Number BRADYLINK WARNING CHAIN WHITE 25Mx8MM
Color White
Material Polyethylene
Description Brady Warning Chain
Dimensions (L x W x H) 25 m x 28 mm x 50 mm
For Use With (Application) Traffic Control
Height (English) 1.97 in.
Height (Metric) 50 mm
Length (English) 82 ft.
Length (Metric) 25 m
Quantity 1 Ea.
Width (English) 1.1 in.
Width (Metric) 28 mm