Learn More
Köttermann Wall-mounted Cabinet
Create additional storage space in any laboratory with this Wall-mounted Cabinet.
Brand: Köttermann 307-00063
Additional Details : Weight : 23.95560kg
Description
- Unobtrusive storage space
- Features lockable door
- Constructed of durable epoxy coated steel
Specifications
366 x 600 x 780 mm | |
None | |
1 | |
Epoxy Coated | |
1 | |
Steel |
Right | |
Hinged | |
Wall Mounted Cabinet | |
Yes | |
White |