Learn More
Sartorius Arium™ pro Ultrapure Water Systems
BIOPHARMA
Product Code. 11725016 Shop All Sartorius Products
Change view
Click to view available options
Bacterial Content:
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bacterial Content Unit Size
11725016 <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11725016 Supplier Sartorius Supplier No. H2OPROVFB
  • ONLINE EXCLUSIVE PRICE
    118900.00 NOK
    70201.11 NOK / Pack of 1
    Save 48698.89 NOK
    40% Off
 Restricted Item
In Stock
Intuitive menu navigation ensures error-free operation in consistent water quantity.
  • pro VF Wall-Mounted System
  • High Efficiency UV Lamp (185/254 nm) and TOC Monitoring
  • Eliminates bacteria and organics
  • Display with touch-function
  • Integrated real-time monitoring
  • Includes wall bracket for the system, UV Lamp and Ultrafilter
TRUSTED_SUSTAINABILITY
Specifications
Description Ultrapure Water Systems
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Distilled, DI, RO, EDI
Flow Rate 1.7 L/min
Product Water Type Type 1
Remote Dispenser No
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring No
Total Organic Carbon ≤2 ppb
Includes wall bracket for the system, UV Lamp and Ultrafilter
Mounting Wall
Daily Output 100 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Arium pro Ultrapure Water System