Learn More
Kinematica W-Design Aggregate with Knives for PT 2500 E
Product Code. 15797198 Shop All Kinematica Products
Change view
Click to view available options
Speed:
21 m/s
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
15797198 21 m/s Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15797198 Supplier Kinematica Supplier No. 11030276
  • ONLINE EXCLUSIVE PRICE
    28160.00 NOK
    26614.72 NOK / Pack of 1
    Save 1545.28 NOK
    5% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Prevents fibrous, stringy, or solid samples from clogging the aggregate.
  • Selection and applications:
    • Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
    • All standard dispersing applications similar to W- or M-EC aggregate
Specifications
Speed 21 m/s
Volume Range (Metric) 100 mL to 2.5 L
Diameter (Metric) Probe 20 mm
Length (Metric) 192 mm
Packaging Quantity 1 Ea.
Product Type W-design, with knives