Learn More
VX-984, MedChemExpress
Product Code. 16417978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16417978 10 mg 10mg
16450438 5 mg 5mg
16403907 50 mg 50mg
3 options
Product Code. 16417978
Brand: Medchem Express HY19939S10mg
MedChemExpress VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.
VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research.
Specifications
Chemical Name or Material VX-984
CAS 1476074-39-1
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C23H21D2N7O
Color White
Synonym M9831
Solubility Information DMSO : 10 mg/mL (24.07 mM; Need ultrasonic)
SMILES O=C(C1=CC=NC2=C([C@H](C)CNC3=CC(C4=C([2H])N=C(C)N=C4[2H])=NC=N3)C=CC=C12)NC
Industry Type Cancer-Kinase/protease
Molecular Weight (g/mol) 415.49
Quantity 10 mg
Formula Weight 415.49
Percent Purity 98.34%
Grade Research