Learn More
VX 745, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(2,6-Dichlorophenyl)-2-[(2,4-difluorophenyl)thio]-6H-pyrimido[1,6-b]pyridazin-6-one |
| CAS | 209410-46-8 |
| Molecular Formula | C19H9Cl2F2N3OS |
| Quantity | 25 mg |
| Synonym | VX 745, 5-(2,6-Dichlorophenyl)-2-[(2,4-difluorophenyl)thio]-6H-pyrimido[1,6-b]pyridazin-6-one |
| InChI Formula | InChI=1S/C19H9Cl2F2N3OS/c20-11-2-1-3-12(21)17(11)18-14-5-7-16(25-26(14)9-24-19(18)27)28-15-6-4-10(22)8-13(15)23/h1-9H |
| SMILES | Fc1ccc(SC2=NN3C=NC(=O)C(=C3C=C2)c4c(Cl)cccc4Cl)c(F)c1 |
| IUPAC Name | 5-(2,6-dichlorophenyl)-2-(2,4-difluorophenyl)sulfanylpyrimido[1,6-b]pyridazin-6-one |
| Molecular Weight (g/mol) | 436.26 |
| Formula Weight | 434.9811 |