missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWF Polyclonal Antibody

Description
PA1-43057 detects von Willebrand Factor in human samples. PA1-43057 has successfully been used in Western blot and ELISA applications. PA1-43057 was derived from human von Willebrand factor.
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
Specifications
Specifications
| Antigen | VWF |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | VWF |
| Gene Accession No. | P04275 |
| Gene Alias | 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 |
| Gene Symbols | VWF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?