Learn More
VU6012962, MedChemExpress
Product Code. 16495616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16495616 100 mg 100mg
16451087 50 mg 50mg
16444828 10 mg 10mg
16437577 5 mg 5mg
4 options
Product Code. 16495616
Brand: Medchem Express HY114403
MedChemExpress VU6012962 is an orally bioavailable and CNS-penetrant metabotropic glutamate receptor 7 negative allosteric modulator (mGlu7 NAM) with an IC50 of 347 nM.
VU6012962 is an orally bioavailable and CNS-penetrant metabotropic glutamate receptor 7 negative allosteric modulator (mGlu7 NAM) with an IC50 of 347 nM.
Specifications
Chemical Name or Material VU6012962
CAS 2313526-86-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H19F3N4O4
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (278.77 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OCC2CC2)C(OC)=C1)NC3=C(N4C=NC=N4)C=CC(OC(F)(F)F)=C3
Molecular Weight (g/mol) 448.4
Formula Weight 448.4
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation