Learn More
VU0152100, MedChemExpress
Product Code. 16465658
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16465658
Brand: Medchem Express HY13340
MedChemExpress VU0152100 is a potent and selective allosteric potentiator of M4 mAChR with an EC50 of 380 (plusmn) 93 nM.
VU0152100 is a potent and selective allosteric potentiator of M4 mAChR with an EC50 of 380 (plusmn) 93 nM.
Specifications
Chemical Name or Material VU0152100
CAS 409351-28-6
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C18H19N3O2S
Quantity 10 mg
Synonym VU152100
Solubility Information DMSO : ≥ 50 mg/mL (146.44 mM)
SMILES CC1=C2C(SC(C(NCC3=CC=C(OC)C=C3)=O)=C2N)=NC(C)=C1
Molecular Weight (g/mol) 341.43
Formula Weight 341.43
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.