missing translation for 'onlineSavingsMsg'
Learn More

VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech

Product Code. 16898235 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16898235 20 μL 20µL
16888235 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16898235

Brand: Proteintech 663981IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.

Specifications

Antigen VTN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D2C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VTN
Gene Accession No. P04004, P22458, P48819
Gene Alias S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN
Gene Symbols VTN
Host Species Mouse
Immunogen VTN Fusion Protein Ag8443
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009128, 397192, 7448
Target Species Rabbit, Pig, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.