missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Antibody (2174B) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB46461JF525
4199.00 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Flow Cytometry,Western Blot
Specifications
| VSIG4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Mouse myeloma cell line NS0-derived human VSIG4, Arg20-Pro283, Accession # Q9Y279 | |
| RUO | |
| 11326 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Western Blot | |
| 2174B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction