missing translation for 'onlineSavingsMsg'
Learn More

Vps34-IN-2, MedChemExpress

Product Code. p-200100051
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471259 1 mg 1mg
16457917 5 mg 5mg
16405198 10 mg 10mg
16403316 25 mg 25mg
16401417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471259 Supplier Medchem Express Supplier No. HY12473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43.

Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43.

Specifications

Chemical Name or Material Vps34-IN-2
Physical Form Solid
CAS 1523404-29-6
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C18H25F3N4O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information Ethanol : 50 mg/mL (124.25 mM; Need ultrasonic)
SMILES O=C1C=C(N2[C@H](C)COCC2)N=C3N(CC(C(C)C)=O)[C@H](C(F)(F)F)CCN31
Molecular Weight (g/mol) 402.41
Formula Weight 402.41
Percent Purity 99.49%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.