Learn More
VPS33A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16896-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16886654
  • 4593.76 NOK / 150µL
Estimated Shipment: 22-05-2024
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predomitly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifications
Specifications
VPS33A
Polyclonal
Unconjugated
VPS33A
hVPS33A, VPS33A
Rabbit
Antigen Affinity Chromatography
RUO
65081, 65082, 77573
-20°C
Liquid
Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q63615, Q96AX1, Q9D2N9
Vps33a
VPS33A Fusion Protein Ag10422
150 μL
Primary
Human, Mouse, Rat