missing translation for 'onlineSavingsMsg'
Learn More

Voxtalisib, MedChemExpress

Product Code. p-200096416
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496908

Brand: Medchem Express HY15900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Voxtalisib (XL765) is a potent PI3K inhibitor, which has a similar activity toward class I PI3K (IC50s=39, 113, 9 and 43 nM for p110α, p110β, p110γ and p110δ, respectively), also inhibits DNA-PK (IC50=150 nM) and mTOR (IC50=157 nM). Voxtalisib (XL765) inhibits mTORC1 and mTORC2 with IC50s of 160 and 910 nM, respectively.

Voxtalisib (XL765) is a potent PI3K inhibitor, which has a similar activity toward class I PI3K (IC50s=39, 113, 9 and 43 nM for p110α, p110β, p110γ and p110δ, respectively), also inhibits DNA-PK (IC50=150 nM) and mTOR (IC50=157 nM). Voxtalisib (XL765) inhibits mTORC1 and mTORC2 with IC50s of 160 and 910 nM, respectively.

Specifications

Chemical Name or Material Voxtalisib
Physical Form Solid
CAS 934493-76-2
Color Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H14N6O
For Use With (Application) Cancer-Kinase/protease
Synonym XL765 SAR245409
Solubility Information DMSO : 10 mg/mL (37.00 mM; Need ultrasonic)
SMILES O=C1C(C2=NNC=C2)=CC3=C(C)N=C(N)N=C3N1CC
Molecular Weight (g/mol) 270.29
Formula Weight 270.29
Percent Purity 98.68%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.