missing translation for 'onlineSavingsMsg'
Learn More

Vortioxetine hydrobromide, MedChemExpress

Product Code. p-200099012
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16466498 10 mg 10mg
16452597 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 16466498

Brand: Medchem Express HY15414A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vortioxetine hydrobromide is a multimodal serotonergic agent, inhibits 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively.

Vortioxetine hydrobromide is a multimodal serotonergic agent, inhibits 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively.

Specifications

Chemical Name or Material Vortioxetine hydrobromide
Physical Form Solid
CAS 960203-27-4
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H23BrN2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Lu AA21004 hydrobromide
Solubility Information DMSO : 50 mg/mL (131.80 mM; Need ultrasonic)
SMILES CC1=CC=C(SC2=CC=CC=C2N3CCNCC3)C(C)=C1.[H]Br
Molecular Weight (g/mol) 379.36
Formula Weight 379.36
Percent Purity 99.45%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.